ContactUsLinkComponent
Rat TGF beta 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of rat TGF beta 2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 96 Tests
Catalog # ANTIA2537-96
Supplier:  ANTIBODIES.COM
New Product
Rat TGF beta 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of rat TGF beta 2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 96 Tests
Catalog # ANTIA2537-96
Supplier:  ANTIBODIES.COM