ContactUsLinkComponent
Rat Hydrogen Potassium ATPase beta/ATP4B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitr o quantitative determination of rat Hydrogen Potassium ATPase beta/ATP4B in tissue homogenates or other biological fl 1 * 48 Tests
Catalog # ANTIA270970-48
Supplier:  ANTIBODIES.COM
New Product
Rat Hydrogen Potassium ATPase beta/ATP4B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitr o quantitative determination of rat Hydrogen Potassium ATPase beta/ATP4B in tissue homogenates or other biological fl 1 * 48 Tests
Catalog # ANTIA270970-48
Supplier:  ANTIBODIES.COM