Porcine HSP90 alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of porcine HSP90 alpha in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other b 1 * 96 Tests
Catalog # ANTIA270958-96
Supplier: Antibodies.com