ContactUsLinkComponent
Mouse PDP1/PDP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of mouse PDP1/PDP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 96 Tests
Catalog # ANTIA1998-96
Supplier:  Antibodies.com
New Product
Mouse PDP1/PDP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of mouse PDP1/PDP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 96 Tests
Catalog # ANTIA1998-96
Supplier:  Antibodies.com