ContactUsLinkComponent
Mouse Myosin Light Chain Kinase/MLCK ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of mouse Myosin Light Chain Kinase/MLCK in tissue homogenates, cell lysates or other biologi 1 * 48 Tests
Catalog # ANTIA270678-48
Supplier:  ANTIBODIES.COM
New Product
Mouse Myosin Light Chain Kinase/MLCK ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of mouse Myosin Light Chain Kinase/MLCK in tissue homogenates, cell lysates or other biologi 1 * 48 Tests
Catalog # ANTIA270678-48
Supplier:  ANTIBODIES.COM