ContactUsLinkComponent
Mouse Complement C9 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of mouse Complement C9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other b 1 * 48 Tests
Catalog # ANTIA270643-48
Supplier:  Antibodies.com
New Product
Mouse Complement C9 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of mouse Complement C9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other b 1 * 48 Tests
Catalog # ANTIA270643-48
Supplier:  Antibodies.com