ContactUsLinkComponent
Human Cross Linked C-Telopeptide Of Type III Collagen ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed fo r the in vitro quantitative determination of human Cross Linked C-Telopeptide Of Type III Collagen in serum, plasma o 1 * 48 Tests
Catalog # ANTIA1869-48
Supplier:  ANTIBODIES.COM
New Product
Human Cross Linked C-Telopeptide Of Type III Collagen ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed fo r the in vitro quantitative determination of human Cross Linked C-Telopeptide Of Type III Collagen in serum, plasma o 1 * 48 Tests
Catalog # ANTIA1869-48
Supplier:  ANTIBODIES.COM