ContactUsLinkComponent
Human Peptide YY/PYY ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative de termination of human Peptide YY/PYY in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or ot 1 * 48 Tests
Catalog # ANTIA2050-48
Supplier:  ANTIBODIES.COM
New Product
Human Peptide YY/PYY ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative de termination of human Peptide YY/PYY in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or ot 1 * 48 Tests
Catalog # ANTIA2050-48
Supplier:  ANTIBODIES.COM