ContactUsLinkComponent
Human Carbohydrate Antigen 72-4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Carbohydrate Antigen 72-4 in serum, plasma, tissue homogenates, cell lysates, cell cultu 1 * 96 Tests
Catalog # ANTIA2367-96
Supplier:  ANTIBODIES.COM
New Product
Human Carbohydrate Antigen 72-4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Carbohydrate Antigen 72-4 in serum, plasma, tissue homogenates, cell lysates, cell cultu 1 * 96 Tests
Catalog # ANTIA2367-96
Supplier:  ANTIBODIES.COM