ContactUsLinkComponent
Human Lipocalin-2/NGAL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of human Lipocalin-2/NGAL in serum, plasma, urine, tissue homogenates, cell lysates, cell culture supernat 1 * 48 Tests
Catalog # ANTIA2066-48
Supplier:  ANTIBODIES.COM
New Product
Human Lipocalin-2/NGAL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of human Lipocalin-2/NGAL in serum, plasma, urine, tissue homogenates, cell lysates, cell culture supernat 1 * 48 Tests
Catalog # ANTIA2066-48
Supplier:  ANTIBODIES.COM