ContactUsLinkComponent
Mouse Myosin Heavy Chain 1/MYH1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of mouse Myosin Heavy Chain 1/MYH1 in serum, plasma, tissue homogenates or other biological fluid 1 * 48 Tests
Catalog # ANTIA2679-48
Supplier:  ANTIBODIES.COM
New Product
Mouse Myosin Heavy Chain 1/MYH1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of mouse Myosin Heavy Chain 1/MYH1 in serum, plasma, tissue homogenates or other biological fluid 1 * 48 Tests
Catalog # ANTIA2679-48
Supplier:  ANTIBODIES.COM