ContactUsLinkComponent
Human MIP-1 alpha/CCL3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of human MIP-1 alpha/CCL3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or o 1 * 48 Tests
Catalog # ANTIA2804-48
Supplier:  ANTIBODIES.COM
New Product
Human MIP-1 alpha/CCL3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of human MIP-1 alpha/CCL3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or o 1 * 48 Tests
Catalog # ANTIA2804-48
Supplier:  ANTIBODIES.COM