Human Mid-regonal pro-Adrenomedullin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Mid-regonal pro-Adrenomedullin in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA246921-96
Supplier: Antibodies.com