Human alpha 1 Acid Glycoprotein/AGP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of human alpha 1 Acid Glycoprotein/AGP in serum, plasma, tissue homogenates, cell lysates, ce 1 * 48 Tests
Catalog # ANTIA1970-48
Supplier: Antibodies.com