ContactUsLinkComponent
Porcine TGF beta 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of porcine TGF beta 3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other bio 1 * 96 Tests
Catalog # ANTIA2775-96
Supplier:  ANTIBODIES.COM
New Product
Porcine TGF beta 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of porcine TGF beta 3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other bio 1 * 96 Tests
Catalog # ANTIA2775-96
Supplier:  ANTIBODIES.COM