ContactUsLinkComponent
Bovine IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antib ody, Streptavidin-HRP, recombinant bovine IL-17A ELISA standard, and standard reconstitution buffer. This matched ant 1 * 1.920 Tests
Catalog # ANTIA270459-2096
Supplier:  ANTIBODIES.COM
New Product
Bovine IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antib ody, Streptavidin-HRP, recombinant bovine IL-17A ELISA standard, and standard reconstitution buffer. This matched ant 1 * 1.920 Tests
Catalog # ANTIA270459-2096
Supplier:  ANTIBODIES.COM