ContactUsLinkComponent
Human IL-1 alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-AP, recombinant human IL-1a ELISA standard, and standard reconstitution buffer. This matched ant 1 * 1.920 Tests
Catalog # ANTIA270398-2096
Supplier:  ANTIBODIES.COM
New Product
Human IL-1 alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-AP, recombinant human IL-1a ELISA standard, and standard reconstitution buffer. This matched ant 1 * 1.920 Tests
Catalog # ANTIA270398-2096
Supplier:  ANTIBODIES.COM