ContactUsLinkComponent
Mouse Complement C3a ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of mouse Complement C3a in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid 1 * 96 Tests
Catalog # ANTIA326437-96
Supplier:  Antibodies.com
New Product
Mouse Complement C3a ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of mouse Complement C3a in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid 1 * 96 Tests
Catalog # ANTIA326437-96
Supplier:  Antibodies.com