ContactUsLinkComponent
Mouse Apelin 36 ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determi nation of mouse Apelin 36 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samples 1 * 96 Tests
  ANTIA326404-96
New Product
Mouse Apelin 36 ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determi nation of mouse Apelin 36 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samples 1 * 96 Tests
  ANTIA326404-96