ContactUsLinkComponent
Mouse IL22 RA2/IL-22BP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of mouse IL22 RA2/IL-22BP in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liq 1 * 96 Tests
Catalog # ANTIA326658-96
Supplier:  ANTIBODIES.COM
New Product
Mouse IL22 RA2/IL-22BP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of mouse IL22 RA2/IL-22BP in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liq 1 * 96 Tests
Catalog # ANTIA326658-96
Supplier:  ANTIBODIES.COM