ContactUsLinkComponent
Rat PI 3 Kinase Catalytic Subunit alpha/PIK3CA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the i n vitro quantitative determination of rat PI 3 Kinase Catalytic Subunit alpha/PIK3CA in serum, plasma, tissue homogen 1 * 96 Tests
Catalog # ANTIA303701-96
Supplier:  ANTIBODIES.COM
New Product
Rat PI 3 Kinase Catalytic Subunit alpha/PIK3CA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the i n vitro quantitative determination of rat PI 3 Kinase Catalytic Subunit alpha/PIK3CA in serum, plasma, tissue homogen 1 * 96 Tests
Catalog # ANTIA303701-96
Supplier:  ANTIBODIES.COM