ContactUsLinkComponent
Human Anti-Myosin Heavy Chain/MHC Antibody ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quant itative determination of human Anti-Myosin Heavy Chain/MHC Antibody in serum, plasma, tissue homogenates, and other b 1 * 96 Tests
Catalog # ANTIA303117-96
Supplier:  ANTIBODIES.COM
New Product
Human Anti-Myosin Heavy Chain/MHC Antibody ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quant itative determination of human Anti-Myosin Heavy Chain/MHC Antibody in serum, plasma, tissue homogenates, and other b 1 * 96 Tests
Catalog # ANTIA303117-96
Supplier:  ANTIBODIES.COM