ContactUsLinkComponent
Human Epstein Barr Virus Early Antigen IgG ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quali tative determination of human Epstein Barr Virus Early Antigen IgG in serum, plasma, tissue homogenates, and other bi 1 * 96 Tests
Catalog # ANTIA303092-96
Supplier:  ANTIBODIES.COM
New Product
Human Epstein Barr Virus Early Antigen IgG ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quali tative determination of human Epstein Barr Virus Early Antigen IgG in serum, plasma, tissue homogenates, and other bi 1 * 96 Tests
Catalog # ANTIA303092-96
Supplier:  ANTIBODIES.COM