ContactUsLinkComponent
Human Troponin I Fast Skeletal Muscle ELISA Kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Troponin I Fast Skeletal Muscle in serum, plasma, and other biological fl 1 * 96 Tests
Catalog # ANTIA312588-96
Supplier:  Antibodies.com
New Product
Human Troponin I Fast Skeletal Muscle ELISA Kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Troponin I Fast Skeletal Muscle in serum, plasma, and other biological fl 1 * 96 Tests
Catalog # ANTIA312588-96
Supplier:  Antibodies.com