ContactUsLinkComponent
Human Anti-SARS-CoV-2 Spike RBD (omicron BA.5.2) IgG ELISA Kit is an indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of human Anti-SARS-CoV-2 Spike RBD (omicron BA.5.2) IgG in serum, plasma, tissue homo 1 * 96 Tests
Catalog # ANTIA319525-96
Supplier:  ANTIBODIES.COM
New Product
Human Anti-SARS-CoV-2 Spike RBD (omicron BA.5.2) IgG ELISA Kit is an indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of human Anti-SARS-CoV-2 Spike RBD (omicron BA.5.2) IgG in serum, plasma, tissue homo 1 * 96 Tests
Catalog # ANTIA319525-96
Supplier:  ANTIBODIES.COM