ContactUsLinkComponent
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Claudin 9 protein to be purified and analysed in vitro. 1 * 50 µG
  ANTIA318429-50
New Product
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Claudin 9 protein to be purified and analysed in vitro. 1 * 50 µG
  ANTIA318429-50