Human Cytokeratin 15 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Cytokeratin 15 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid 1 * 96 Tests
Catalog # ANTIA326520-96
Supplier: ANTIBODIES.COM