ContactUsLinkComponent
Canine Complement C5a ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of canine Complement C5a in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liqui 1 * 96 Tests
Catalog # ANTIA326445-96
Supplier:  ANTIBODIES.COM
New Product
Canine Complement C5a ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of canine Complement C5a in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liqui 1 * 96 Tests
Catalog # ANTIA326445-96
Supplier:  ANTIBODIES.COM