ContactUsLinkComponent
Porcine IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection anti body, Streptavidin-AP, recombinant bovine IL-17A ELISA standard, and standard reconstitution buffer. This matched ant 1 * 576 Tests
Catalog # ANTIA325370-696
Supplier:  ANTIBODIES.COM
New Product
Porcine IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection anti body, Streptavidin-AP, recombinant bovine IL-17A ELISA standard, and standard reconstitution buffer. This matched ant 1 * 576 Tests
Catalog # ANTIA325370-696
Supplier:  ANTIBODIES.COM