ContactUsLinkComponent
Human SERPINB1/PI2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of human SERPINB1/PI2 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samp 1 * 96 Tests
Catalog # ANTIA326882-96
Supplier:  ANTIBODIES.COM
New Product
Human SERPINB1/PI2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of human SERPINB1/PI2 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samp 1 * 96 Tests
Catalog # ANTIA326882-96
Supplier:  ANTIBODIES.COM