Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human PAR1/Thrombin Receptor protein to be purified and analysed in vitro. 1 * 10 µG
Catalog # ANTIA325277-10
Supplier: ANTIBODIES.COM