ContactUsLinkComponent
Mouse Creatine Kinase MM ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of mouse Creatine Kinase MM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 96 Tests
Catalog # ANTIA3044-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Creatine Kinase MM ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of mouse Creatine Kinase MM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 96 Tests
Catalog # ANTIA3044-96
Supplier:  ANTIBODIES.COM