ContactUsLinkComponent
Mouse Leukotriene B4 ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative de termination of mouse Leukotriene B4 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liqu 1 * 96 Tests
  ANTIA326715-96
New Product
Mouse Leukotriene B4 ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative de termination of mouse Leukotriene B4 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liqu 1 * 96 Tests
  ANTIA326715-96