ContactUsLinkComponent
Mouse alpha 1,2 Mannosidase IA/MAN1A1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro q uantitative determination of mouse alpha 1,2 Mannosidase IA/MAN1A1 in serum, plasma, tissue homogenates, and other bi 1 * 96 Tests
Catalog # ANTIA319393-96
Supplier:  ANTIBODIES.COM
New Product
Mouse alpha 1,2 Mannosidase IA/MAN1A1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro q uantitative determination of mouse alpha 1,2 Mannosidase IA/MAN1A1 in serum, plasma, tissue homogenates, and other bi 1 * 96 Tests
Catalog # ANTIA319393-96
Supplier:  ANTIBODIES.COM