ContactUsLinkComponent
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human GABA B Receptor 2/GABBR2 protein to be purified and analysed in vitro. 1 * 50 µG
Catalog # ANTIA325143-50
Supplier:  ANTIBODIES.COM
New Product
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human GABA B Receptor 2/GABBR2 protein to be purified and analysed in vitro. 1 * 50 µG
Catalog # ANTIA325143-50
Supplier:  ANTIBODIES.COM