ContactUsLinkComponent
Human Niemann Pick C2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of human Niemann Pick C2 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liqui 1 * 96 Tests
Catalog # ANTIA326768-96
Supplier:  ANTIBODIES.COM
New Product
Human Niemann Pick C2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of human Niemann Pick C2 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liqui 1 * 96 Tests
Catalog # ANTIA326768-96
Supplier:  ANTIBODIES.COM