ContactUsLinkComponent
Mouse HMW Kininogen ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of mouse HMW Kininogen in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid sa 1 * 96 Tests
  ANTIA326695-96
New Product
Mouse HMW Kininogen ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of mouse HMW Kininogen in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid sa 1 * 96 Tests
  ANTIA326695-96