ContactUsLinkComponent
Guinea Pig Complement C4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of guinea pig Complement C4 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other 1 * 96 Tests
Catalog # ANTIA326441-96
Supplier:  ANTIBODIES.COM
New Product
Guinea Pig Complement C4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of guinea pig Complement C4 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other 1 * 96 Tests
Catalog # ANTIA326441-96
Supplier:  ANTIBODIES.COM