Human 25-Hydroxy Vitamin D ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitat ive determination of human 25-Hydroxy Vitamin D in serum, plasma, cell culture supernatant, cell or tissue lysate, an 1 * 96 Tests
Catalog # ANTIA326317-96
Supplier: ANTIBODIES.COM