ContactUsLinkComponent
Guinea Pig Mast Cell Tryptase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of guinea pig Mast Cell Tryptase in serum, plasma, cell culture supernatant, cell or tissue lysate, 1 * 96 Tests
Catalog # ANTIA326932-96
Supplier:  ANTIBODIES.COM
New Product
Guinea Pig Mast Cell Tryptase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of guinea pig Mast Cell Tryptase in serum, plasma, cell culture supernatant, cell or tissue lysate, 1 * 96 Tests
Catalog # ANTIA326932-96
Supplier:  ANTIBODIES.COM