ContactUsLinkComponent
Human Anti-SARS-CoV-2 Spike RBD (omicron B.1.1.529) IgM ELISA Kit is an indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of human Anti-SARS-CoV-2 Spike RBD (omicron B.1.1.529) IgM in serum, plasma, tissu 1 * 96 Tests
Catalog # ANTIA319451-96
Supplier:  ANTIBODIES.COM
New Product
Human Anti-SARS-CoV-2 Spike RBD (omicron B.1.1.529) IgM ELISA Kit is an indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of human Anti-SARS-CoV-2 Spike RBD (omicron B.1.1.529) IgM in serum, plasma, tissu 1 * 96 Tests
Catalog # ANTIA319451-96
Supplier:  ANTIBODIES.COM