ContactUsLinkComponent
Human Dystrophia Myotonica Protein Kinase / DMPK ELISA Kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) design ed for the in vitro quantitative determination of human Dystrophia Myotonica Protein Kinase / DMPK in serum, plasma, 1 * 96 Tests
Catalog # ANTIA311128-96
Supplier:  ANTIBODIES.COM
New Product
Human Dystrophia Myotonica Protein Kinase / DMPK ELISA Kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) design ed for the in vitro quantitative determination of human Dystrophia Myotonica Protein Kinase / DMPK in serum, plasma, 1 * 96 Tests
Catalog # ANTIA311128-96
Supplier:  ANTIBODIES.COM