ContactUsLinkComponent
Human Semaphorin 3C ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of human Semaphorin 3C in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid sa 1 * 96 Tests
Catalog # ANTIA326881-96
Supplier:  ANTIBODIES.COM
New Product
Human Semaphorin 3C ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of human Semaphorin 3C in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid sa 1 * 96 Tests
Catalog # ANTIA326881-96
Supplier:  ANTIBODIES.COM