ContactUsLinkComponent
Guinea Pig CXCL1 (GRO alpha) ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitati ve determination of guinea pig CXCL1 (GRO alpha) in serum, plasma, cell culture supernatant, cell or tissue lysate, a 1 * 96 Tests
Catalog # ANTIA326511-96
Supplier:  ANTIBODIES.COM
New Product
Guinea Pig CXCL1 (GRO alpha) ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitati ve determination of guinea pig CXCL1 (GRO alpha) in serum, plasma, cell culture supernatant, cell or tissue lysate, a 1 * 96 Tests
Catalog # ANTIA326511-96
Supplier:  ANTIBODIES.COM