ContactUsLinkComponent
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length, native-folded transmembrane human Adenine Nucleotide Translocator 1/ANT1 protein to be purified and analysed in vitr 1 * 10 µG
Catalog # ANTIA318466-10
Supplier:  ANTIBODIES.COM
New Product
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length, native-folded transmembrane human Adenine Nucleotide Translocator 1/ANT1 protein to be purified and analysed in vitr 1 * 10 µG
Catalog # ANTIA318466-10
Supplier:  ANTIBODIES.COM