ContactUsLinkComponent
Human Sodium Iodide Symporter ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of human Sodium Iodide Symporter in serum, plasma, cell culture supernatant, cell or tissue lysate, 1 * 96 Tests
Catalog # ANTIA326761-96
Supplier:  ANTIBODIES.COM
New Product
Human Sodium Iodide Symporter ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of human Sodium Iodide Symporter in serum, plasma, cell culture supernatant, cell or tissue lysate, 1 * 96 Tests
Catalog # ANTIA326761-96
Supplier:  ANTIBODIES.COM