ContactUsLinkComponent
Mouse Myeloid-derived Growth Factor/SF20 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitr o quantitative determination of mouse Myeloid-derived Growth Factor/SF20 in serum, plasma, tissue homogenates, and ot 1 * 96 Tests
Catalog # ANTIA303539-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Myeloid-derived Growth Factor/SF20 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitr o quantitative determination of mouse Myeloid-derived Growth Factor/SF20 in serum, plasma, tissue homogenates, and ot 1 * 96 Tests
Catalog # ANTIA303539-96
Supplier:  ANTIBODIES.COM