ContactUsLinkComponent
Mouse Anti-SARS-CoV-2 Spike RBD Antibody ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantit ative determination of mouse Anti-SARS-CoV-2 Spike RBD Antibody in serum, plasma, cell culture supernatant, cell or t 1 * 96 Tests
Catalog # ANTIA326398-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Anti-SARS-CoV-2 Spike RBD Antibody ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantit ative determination of mouse Anti-SARS-CoV-2 Spike RBD Antibody in serum, plasma, cell culture supernatant, cell or t 1 * 96 Tests
Catalog # ANTIA326398-96
Supplier:  ANTIBODIES.COM