ContactUsLinkComponent
This Hydroxyl Radical Antioxidant Capacity Fluorometric Assay Kit is a quantitative assay kit for measuring Hydroxyl Radical Antioxi dant Capacity (HORAC) activity in serum, plasma, animal and plant tissues. 1 * 48 Tests
Catalog # ANTIA319648-48
Supplier:  ANTIBODIES.COM
New Product
This Hydroxyl Radical Antioxidant Capacity Fluorometric Assay Kit is a quantitative assay kit for measuring Hydroxyl Radical Antioxi dant Capacity (HORAC) activity in serum, plasma, animal and plant tissues. 1 * 48 Tests
Catalog # ANTIA319648-48
Supplier:  ANTIBODIES.COM