ContactUsLinkComponent
Mouse IL-1 beta Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection ant ibody, Streptavidin-AP, recombinant mouse IL-1&#0223 ELISA standard, and standard reconstitution buffer. This matche 1 * 576 Tests
Catalog # ANTIA325338-696
Supplier:  ANTIBODIES.COM
New Product
Mouse IL-1 beta Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection ant ibody, Streptavidin-AP, recombinant mouse IL-1&#0223 ELISA standard, and standard reconstitution buffer. This matche 1 * 576 Tests
Catalog # ANTIA325338-696
Supplier:  ANTIBODIES.COM